Gene Transcripts
- Gene Symbol
- Txndc5
- Entrez Gene ID
- 105245
- Gene Name
- thioredoxin domain containing 5
- Gene Aliases
- AL022641, ERp46, PC-TRP
- Chromosome Location
- Chr. 13: 38500266 - 38528824 on Build GRCm38
- UniGene ID
- Mm.28622
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289598.1 |
NP_001276527.1
|
4, 5 |
634 |
|
NM_001289599.1 |
NP_001276528.1
|
4, 5 |
601 |
|
NM_145367.4 |
NP_663342.3
|
4, 5 |
658 |
|
GenBank mRNA |
AK136046.1 |
- |
4, 5 |
546 |
|
AK142049.1 |
- |
4, 5 |
644 |
|
AK150781.1 |
- |
4, 5 |
642 |
|
AK169680.1 |
- |
4, 5 |
625 |
|
AK224911.1 |
- |
3, 4 |
311 |
|
AK224913.1 |
- |
4, 5 |
645 |
|
AK224922.1 |
- |
4, 5 |
603 |
|
AK224923.1 |
- |
4, 5 |
625 |
|
AK224924.1 |
- |
4, 5 |
633 |
|
AK224926.1 |
- |
4, 5 |
603 |
|
AK224927.1 |
- |
4, 5 |
654 |
|
AK224930.1 |
- |
4, 5 |
606 |
|
AK224933.1 |
- |
4, 5 |
595 |
|
AK224934.1 |
- |
4, 5 |
649 |
|
AK224936.1 |
- |
4, 5 |
648 |
|
AK224937.1 |
- |
4, 5 |
647 |
|
AK224938.1 |
- |
4, 5 |
642 |
|
AK224939.1 |
- |
4, 5 |
635 |
|
AK224943.1 |
- |
4, 5 |
606 |
|
AK224945.1 |
- |
4, 5 |
642 |
|
AK224946.1 |
- |
4, 5 |
645 |
|
AK224947.1 |
- |
4, 5 |
632 |
|
AK224948.1 |
- |
4, 5 |
606 |
|
AK224949.1 |
- |
4, 5 |
595 |
|
AK224950.1 |
- |
4, 5 |
595 |
|
AK224951.1 |
- |
4, 5 |
653 |
|
AK224952.1 |
- |
4, 5 |
621 |
|
AK224956.1 |
- |
4, 5 |
565 |
|
AK224957.1 |
- |
4, 5 |
611 |
|
AK224958.1 |
- |
4, 5 |
611 |
|
AK224960.1 |
- |
4, 5 |
653 |
|
AK224962.1 |
- |
4, 5 |
613 |
|
AK224963.1 |
- |
4, 5 |
654 |
|
AK224964.1 |
- |
4, 5 |
654 |
|
AK224965.1 |
- |
4, 5 |
630 |
|
AK224966.1 |
- |
4, 5 |
630 |
|
AK224968.1 |
- |
2, 3 |
188 |
|
AK224969.1 |
- |
2, 3 |
188 |
|
AK224970.1 |
- |
4, 5 |
595 |
|
AK224971.1 |
- |
4, 5 |
655 |
|
AK224972.1 |
- |
4, 5 |
645 |
|
AK224973.1 |
- |
4, 5 |
595 |
|
AK224974.1 |
- |
4, 5 |
598 |
|
AK224976.1 |
- |
4, 5 |
628 |
|
AK224977.1 |
- |
4, 5 |
642 |
|
AK224978.1 |
- |
4, 5 |
642 |
|
AK224979.1 |
- |
4, 5 |
629 |
|
AK224981.1 |
- |
4, 5 |
599 |
|
AK224982.1 |
- |
4, 5 |
654 |
|
AK224983.1 |
- |
4, 5 |
643 |
|
AK224984.1 |
- |
4, 5 |
608 |
|
AK224985.1 |
- |
4, 5 |
612 |
|
AK224986.1 |
- |
4, 5 |
656 |
|
AK224988.1 |
- |
4, 5 |
632 |
|
AK224989.1 |
- |
4, 5 |
649 |
|
AK224990.1 |
- |
4, 5 |
644 |
|
AK224991.1 |
- |
4, 5 |
639 |
|
AK224992.1 |
- |
4, 5 |
639 |
|
AK224993.1 |
- |
4, 5 |
639 |
|
AK224994.1 |
- |
4, 5 |
647 |
|
AK224995.1 |
- |
4, 5 |
610 |
|
AK224996.1 |
- |
4, 5 |
581 |
|
AK224997.1 |
- |
4, 5 |
606 |
|
AK224998.1 |
- |
4, 5 |
604 |
|
AK224999.1 |
- |
4, 5 |
626 |
|
AK225004.1 |
- |
4, 5 |
647 |
|
AK225005.1 |
- |
4, 5 |
625 |
|
AK225007.1 |
- |
4, 5 |
647 |
|
AK225010.1 |
- |
4, 5 |
647 |
|
AK225011.1 |
- |
4, 5 |
647 |
|
AK225012.1 |
- |
4, 5 |
416 |
|
AK225013.1 |
- |
4, 5 |
416 |
|
AK225014.1 |
- |
4, 5 |
625 |
|
AK225015.1 |
- |
4, 5 |
416 |
|
AK225016.1 |
- |
4, 5 |
555 |
|
AK225017.1 |
- |
4, 5 |
603 |
|
AK225019.1 |
- |
4, 5 |
635 |
|
AK225020.1 |
- |
4, 5 |
586 |
|
AK225021.1 |
- |
4, 5 |
586 |
|
AK225022.1 |
- |
4, 5 |
434 |
|
AK225023.1 |
- |
4, 5 |
660 |
|
AY243534.1 |
- |
4, 5 |
621 |
|
AY548112.1 |
- |
4, 5 |
582 |
|
BC016252.1 |
- |
4, 5 |
625 |
4911455 |
BC024505.1 |
- |
4, 5 |
570 |
4983496 |
BC036155.1 |
- |
3, 4 |
244 |
|
BC046789.1 |
- |
4, 5 |
584 |
5716897 |
BP765107.1 |
- |
4, 5 |
648 |
|
View Full Product Details