Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000856.5 | NP_000847.2 | 7 | 1129 | |
NM_001130682.2 | NP_001124154.1 | 6 | 1035 | ||
NM_001130683.3 | NP_001124155.1 | 6 | 1003 | ||
NM_001130684.2 | NP_001124156.1 | 5 | 975 | ||
NM_001130685.2 | NP_001124157.1 | 6 | 824 | ||
NM_001130687.2 | NP_001124159.1 | 6 | 1035 | ||
NM_001256449.1 | NP_001243378.1 | 6 | 966 | ||
XM_005262955.2 | XP_005263012.1 | 6 | 997 | ||
XM_005262956.2 | XP_005263013.1 | 7 | 812 | ||
XM_005262957.2 | XP_005263014.1 | 7 | 917 | ||
XM_006714196.2 | XP_006714259.1 | 6 | 984 | ||
XM_006714197.2 | XP_006714260.1 | 6 | 759 | ||
XM_006714198.2 | XP_006714261.1 | 6 | 644 | ||
XM_011531900.2 | XP_011530202.1 | 6 | 723 | ||
GenBank mRNA | AK096588.1 | - | 6 | 966 | |
AK226125.1 | - | 6 | 692 | ||
AK309950.1 | - | 6 | 897 | ||
AK312952.1 | - | 6 | 935 | ||
BC028384.2 | - | 5 | 975 | 4792661 | |
BC095395.1 | - | 6 | 926 | ||
BX647200.1 | - | 6 | 802 | ||
BX649180.1 | - | 6 | 756 | ||
U58855.1 | - | 4 | 610 | ||
X66534.1 | - | 7 | 1109 | ||
Y15723.1 | - | 7 | 1116 |