Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252470.1 | NP_001239399.1 | 2 | 363 | |
NM_153679.2 | NP_710146.1 | 2 | 365 | ||
GenBank mRNA | AF320000.1 | - | 2 | 288 | |
AK035790.1 | - | 2 | 329 | ||
AK087747.1 | - | 2 | 237 | ||
BC066155.1 | - | 2 | 287 | 30089333 | |
BC083317.1 | - | 2 | 248 | ||
CN842042.1 | - | 368 |