gRNA Design Details
- Target DNA Sequence
- CGTGCTCATTTTCCATCCGG
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 46744704 - 46744726 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.35/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS45
- Entrez Gene ID
-
377047
- Gene Name
- protease, serine 45
- Gene Aliases
-
TESSP5
- Chromosome Location
-
Chr.3: 46742091 - 46744755 on GRCh38
- UniGene ID
- Hs.667712
- Species
-
Human