gRNA Design Details

Target DNA Sequence
CGTGCTCATTTTCCATCCGG
PAM Sequence
AGG
Target locus
Chr.3: 46744704 - 46744726 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 88.35/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS45
Entrez Gene ID
377047
Gene Name
protease, serine 45
Gene Aliases
TESSP5
Chromosome Location
Chr.3: 46742091 - 46744755 on GRCh38
UniGene ID
Hs.667712
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_199183.2 NP_954652.2 1