gRNA Design Details

Target DNA Sequence
ATTATGCACCCCAAGTACTG
PAM Sequence
GGG
Target locus
Chr.3: 46744030 - 46744052 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.16/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS45
Entrez Gene ID
377047
Gene Name
protease, serine 45
Gene Aliases
TESSP5
Chromosome Location
Chr.3: 46742091 - 46744755 on GRCh38
UniGene ID
Hs.667712
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_199183.2 NP_954652.2 2