gRNA Design Details

Target DNA Sequence
CTCCGCTCTCATAGTAGCCA
PAM Sequence
GGG
Target locus
Chr.15: 66119098 - 66119120 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 91.84/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MEGF11
Entrez Gene ID
84465
Gene Name
multiple EGF like domains 11
Gene Aliases
-
Chromosome Location
Chr.15: 65895296 - 66253737 on GRCh38
UniGene ID
Hs.712886
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_032445.2 NP_115821.2 4
XM_017022670.1 XP_016878159.1 3
XM_017022671.1 XP_016878160.1 3
XM_017022672.1 XP_016878161.1 3
XM_017022673.1 XP_016878162.1 3