gRNA Design Details
- Target DNA Sequence
- GAAATTCCGAGGACTATCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 18121862 - 18121884 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MAST3
- Entrez Gene ID
-
23031
- Gene Name
- microtubule associated serine/threonine kinase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 18097764 - 18151692 on GRCh38
- UniGene ID
- Hs.466184
- Species
-
Human