gRNA Design Details
- Target DNA Sequence
- GAGTGGTTTAAGGCGGAGCT
- PAM Sequence
- TGG
- Target locus
-
Chr.22: 39955837 - 39955859 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GRAP2
- Entrez Gene ID
-
9402
- Gene Name
- GRB2-related adaptor protein 2
- Gene Aliases
-
GADS, GRAP-2, GRB2L, GRBLG, GRID, GRPL, GrbX, Grf40, Mona, P38
- Chromosome Location
-
Chr.22: 39901073 - 39973343 on GRCh38
- UniGene ID
- Hs.517499
- Species
-
Human