gRNA Design Details
- Target DNA Sequence
- GTCGATGACATCTTCAGGCT
- PAM Sequence
- CGG
- Target locus
-
Chr.10: 11866697 - 11866719 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PROSER2
- Entrez Gene ID
-
254427
- Gene Name
- proline and serine rich 2
- Gene Aliases
-
C10orf47
- Chromosome Location
-
Chr.10: 11823341 - 11872277 on GRCh38
- UniGene ID
- Hs.435775
- Species
-
Human