gRNA Design Details
- Target DNA Sequence
- GTTCCGAATCATCCACGAGA
- PAM Sequence
- CGG
- Target locus
-
Chr.21: 41756623 - 41756645 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- RIPK4
- Entrez Gene ID
-
54101
- Gene Name
- receptor interacting serine/threonine kinase 4
- Gene Aliases
-
ANKK2, ANKRD3, DIK, NKRD3, PKK, PPS2, RIP4
- Chromosome Location
-
Chr.21: 41739369 - 41767089 on GRCh38
- UniGene ID
- Hs.517310
- Species
-
Human