gRNA Design Details
- Target DNA Sequence
- AAATCGAAAGGCATGCTCCT
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 36891026 - 36891048 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GRIK3
- Entrez Gene ID
-
2899
- Gene Name
- glutamate ionotropic receptor kainate type subunit 3
- Gene Aliases
-
EAA5, GLR7, GLUR7, GluK3, GluR7a
- Chromosome Location
-
Chr.1: 36795527 - 37034243 on GRCh38
- UniGene ID
- Hs.128848
- Species
-
Human