gRNA Design Details
- Target DNA Sequence
- AGGACAGCACTGACAGCGAG
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 2856180 - 2856202 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS22
- Entrez Gene ID
-
64063
- Gene Name
- protease, serine 22
- Gene Aliases
-
BSSP-4, SP001LA, hBSSP-4
- Chromosome Location
-
Chr.16: 2852727 - 2858630 on GRCh38
- UniGene ID
- Hs.459709
- Species
-
Human