gRNA Design Details
- Target DNA Sequence
- TACCTGTTCTCTGTGCTGCT
- PAM Sequence
- GGG
- Target locus
-
Chr.16: 2855813 - 2855835 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS22
- Entrez Gene ID
-
64063
- Gene Name
- protease, serine 22
- Gene Aliases
-
BSSP-4, SP001LA, hBSSP-4
- Chromosome Location
-
Chr.16: 2852727 - 2858630 on GRCh38
- UniGene ID
- Hs.459709
- Species
-
Human