gRNA Design Details
- Target DNA Sequence
- TAGGCTTCTACCGAATTCGA
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 43245987 - 43246009 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NIM1K
- Entrez Gene ID
-
167359
- Gene Name
- NIM1 serine/threonine protein kinase
- Gene Aliases
-
NIM1
- Chromosome Location
-
Chr.5: 43192054 - 43280850 on GRCh38
- UniGene ID
- Hs.25845
- Species
-
Human