gRNA Design Details
- Target DNA Sequence
- TACCGAATTCGAGGGGAAAT
- PAM Sequence
- CGG
- Target locus
-
Chr.5: 43245995 - 43246017 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
94.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NIM1K
- Entrez Gene ID
-
167359
- Gene Name
- NIM1 serine/threonine protein kinase
- Gene Aliases
-
NIM1
- Chromosome Location
-
Chr.5: 43192054 - 43280850 on GRCh38
- UniGene ID
- Hs.25845
- Species
-
Human