gRNA Design Details
- Target DNA Sequence
- GTGTCAAAGTAGAGCATACC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 148270105 - 148270127 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK13
- Entrez Gene ID
-
153218
- Gene Name
- serine peptidase inhibitor, Kazal type 13 (putative)
- Gene Aliases
-
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3
- Chromosome Location
-
Chr.5: 148268860 - 148286210 on GRCh38
- UniGene ID
- Hs.483771
- Species
-
Human