gRNA Design Details
- Target DNA Sequence
- ACTGGCGACAATGATGCTCT
- PAM Sequence
- CGG
- Target locus
-
Chr.12: 118981965 - 118981987 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.58/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRRM4
- Entrez Gene ID
-
84530
- Gene Name
- serine/arginine repetitive matrix 4
- Gene Aliases
-
KIAA1853, MU-MB-2.76, nSR100
- Chromosome Location
-
Chr.12: 118981495 - 119163051 on GRCh38
- UniGene ID
- Hs.744964
- Species
-
Human