gRNA Design Details
- Target DNA Sequence
- GCGTTTCTGCTGTTGCTCTT
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 5475113 - 5475135 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Efemp2
- Entrez Gene ID
-
58859
- Gene Name
- epidermal growth factor-containing fibulin-like extracellular matrix protein 2
- Gene Aliases
-
0610011K11Rik, Fbln4, MBP1
- Chromosome Location
-
Chr.19: 5473960 - 5481854 on GRCm38
- UniGene ID
- Mm.276367
- Species
-
Mouse