gRNA Design Details
- Target DNA Sequence
- CATTGCTTGGTGCAAAGCCA
- PAM Sequence
- GGG
- Target locus
-
Chr.19: 6903889 - 6903911 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRE1
- Entrez Gene ID
-
2015
- Gene Name
- adhesion G protein-coupled receptor E1
- Gene Aliases
-
EMR1, TM7LN3
- Chromosome Location
-
Chr.19: 6887549 - 6940453 on GRCh38
- UniGene ID
- Hs.2375
- Species
-
Human