gRNA Design Details
- Target DNA Sequence
- CATCGAAGGGTGGGTACTCC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 135852225 - 135852247 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srrm3
- Entrez Gene ID
-
58212
- Gene Name
- serine/arginine repetitive matrix 3
- Gene Aliases
-
2900083I11Rik, AB041661, SRm300-like, Srrm2l
- Chromosome Location
-
Chr.5: 135806861 - 135874773 on GRCm38
- UniGene ID
- Mm.271230
- Species
-
Mouse