gRNA Design Details
- Target DNA Sequence
- ATTCGATGTCCGTGCCCACC
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 45052830 - 45052852 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CLASRP
- Entrez Gene ID
-
11129
- Gene Name
- CLK4 associating serine/arginine rich protein
- Gene Aliases
-
CLASP, SFRS16, SWAP2
- Chromosome Location
-
Chr.19: 45039040 - 45070956 on GRCh38
- UniGene ID
- Hs.466917
- Species
-
Human