gRNA Design Details
- Target DNA Sequence
- GTTCCGGTTCTTGGCGATGG
- PAM Sequence
- TGG
- Target locus
-
Chr.16: 89919440 - 89919462 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.47/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MC1R
- Entrez Gene ID
-
4157
- Gene Name
- melanocortin 1 receptor
- Gene Aliases
-
CMM5, MSH-R, SHEP2
- Chromosome Location
-
Chr.16: 89917879 - 89920977 on GRCh38
- UniGene ID
- Hs.513829
- Species
-
Human