gRNA Design Details
- Target DNA Sequence
- CATTACCAGCAGACTGGAGA
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 3123703 - 3123725 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BPHL
- Entrez Gene ID
-
670
- Gene Name
- biphenyl hydrolase-like (serine hydrolase)
- Gene Aliases
-
BPH-RP, MCNAA, VACVASE
- Chromosome Location
-
Chr.6: 3118376 - 3153198 on GRCh38
- UniGene ID
- Hs.10136
- Species
-
Human