gRNA Design Details
- Target DNA Sequence
- CGCCTGAAAGGTGGTGGAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 120714005 - 120714027 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MEGF9
- Entrez Gene ID
-
1955
- Gene Name
- multiple EGF like domains 9
- Gene Aliases
-
EGFL5
- Chromosome Location
-
Chr.9: 120600918 - 120714487 on GRCh38
- UniGene ID
- Hs.744903
- Species
-
Human