gRNA Design Details
- Target DNA Sequence
- ACCTATTTGTGGCAGTGACC
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 48319537 - 48319559 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.31/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK8
- Entrez Gene ID
-
646424
- Gene Name
- serine peptidase inhibitor, Kazal type 8 (putative)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 48306846 - 48330311 on GRCh38
- UniGene ID
- Hs.257687
- Species
-
Human