gRNA Design Details

Target DNA Sequence
GGCGACGAGCTCACTCTTAT
PAM Sequence
GGG
Target locus
Chr.2: 26184290 - 26184312 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 95.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
Chromosome Location
Chr.2: 26173091 - 26202405 on GRCh38
UniGene ID
Hs.187912
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001168241.1 NP_001161713.1 4