gRNA Design Details

Target DNA Sequence
CCACGTCTCCATCAGGTACT
PAM Sequence
TGG
Target locus
Chr.1: 28859046 - 28859068 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.17/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
OPRD1
Entrez Gene ID
4985
Gene Name
opioid receptor delta 1
Gene Aliases
OPRD
Chromosome Location
Chr.1: 28812142 - 28863696 on GRCh38
UniGene ID
Hs.372
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000911.3 NP_000902.3 2