gRNA Design Details
- Target DNA Sequence
- GAACATATTGTAGTAGTCGA
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 28859103 - 28859125 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
81.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- OPRD1
- Entrez Gene ID
-
4985
- Gene Name
- opioid receptor delta 1
- Gene Aliases
-
OPRD
- Chromosome Location
-
Chr.1: 28812142 - 28863696 on GRCh38
- UniGene ID
- Hs.372
- Species
-
Human