gRNA Design Details
- Target DNA Sequence
- CTACGTCTTCCAGACCCGCA
- PAM Sequence
- AGG
- Target locus
-
Chr.20: 17531293 - 17531315 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BFSP1
- Entrez Gene ID
-
631
- Gene Name
- beaded filament structural protein 1
- Gene Aliases
-
CP115, CP94, CTRCT33, LIFL-H
- Chromosome Location
-
Chr.20: 17493905 - 17569220 on GRCh38
- UniGene ID
- Hs.129702
- Species
-
Human