gRNA Design Details
- Target DNA Sequence
- GAAGCGCACGGATGCCATGT
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 694891 - 694913 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.93/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS57
- Entrez Gene ID
-
400668
- Gene Name
- protease, serine 57
- Gene Aliases
-
NSP4, PRSSL1, UNQ782
- Chromosome Location
-
Chr.19: 685521 - 695461 on GRCh38
- UniGene ID
- Hs.245146
- Species
-
Human