gRNA Design Details
- Target DNA Sequence
- AAGTCCGTCACGCTGACCCA
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 118352407 - 118352429 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS12
- Entrez Gene ID
-
8492
- Gene Name
- protease, serine 12
- Gene Aliases
-
BSSP-3, BSSP3, MRT1
- Chromosome Location
-
Chr.4: 118280038 - 118353042 on GRCh38
- UniGene ID
- Hs.445857
- Species
-
Human