gRNA Design Details
- Target DNA Sequence
- AATCCTTGCGTAGCCCGAAG
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 17843430 - 17843452 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
97.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- JAK3
- Entrez Gene ID
-
3718
- Gene Name
- Janus kinase 3
- Gene Aliases
-
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
- Chromosome Location
-
Chr.19: 17824782 - 17848071 on GRCh38
- UniGene ID
- Hs.515247
- Species
-
Human