gRNA Design Details
- Target DNA Sequence
- TCTCGCTGAGATACGAAGGG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 130855035 - 130855057 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
93.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ABL1
- Entrez Gene ID
-
25
- Gene Name
- ABL proto-oncogene 1, non-receptor tyrosine kinase
- Gene Aliases
-
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
- Chromosome Location
-
Chr.9: 130713881 - 130887675 on GRCh38
- UniGene ID
- Hs.431048
- Species
-
Human