gRNA Design Details

Target DNA Sequence
TGCTGTAGCCAACCGTGTAG
PAM Sequence
AGG
Target locus
Chr.7: 31084763 - 31084785 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 90.25/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADCYAP1R1
Entrez Gene ID
117
Gene Name
ADCYAP receptor type I
Gene Aliases
PAC1, PAC1R, PACAPR, PACAPRI
Chromosome Location
Chr.7: 31052341 - 31114005 on GRCh38
UniGene ID
Hs.377783
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001118.4 NP_001109.2 8
NM_001199635.1 NP_001186564.1 8
NM_001199636.1 NP_001186565.1 8
NM_001199637.1 NP_001186566.1 6
XM_005249618.4 XP_005249675.1 5
XM_006715645.2 XP_006715708.1 8
XM_017011736.1 XP_016867225.1 7
XM_017011737.1 XP_016867226.1 6
XM_017011738.1 XP_016867227.1 5