gRNA Design Details
- Target DNA Sequence
- GAGCGCTAACTATGGTCGGA
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 81907123 - 81907145 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
97.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRL2
- Entrez Gene ID
-
23266
- Gene Name
- adhesion G protein-coupled receptor L2
- Gene Aliases
-
CIRL2, CL2, LEC1, LPHH1, LPHN2
- Chromosome Location
-
Chr.1: 81306155 - 81992738 on GRCh38
- UniGene ID
- Hs.24212
- Species
-
Human