gRNA Design Details
- Target DNA Sequence
- TTGGGGGTGGTCACAGGTCA
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 49016092 - 49016114 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LHB
- Entrez Gene ID
-
3972
- Gene Name
- luteinizing hormone beta polypeptide
- Gene Aliases
-
CGB4, HH23, LSH-B, LSH-beta
- Chromosome Location
-
Chr.19: 49015980 - 49017090 on GRCh38
- UniGene ID
- Hs.154704
- Species
-
Human