gRNA Design Details
- Target DNA Sequence
- TCCAGGAGTCGGGATGGACT
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 49016026 - 49016048 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
81.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LHB
- Entrez Gene ID
-
3972
- Gene Name
- luteinizing hormone beta polypeptide
- Gene Aliases
-
CGB4, HH23, LSH-B, LSH-beta
- Chromosome Location
-
Chr.19: 49015980 - 49017090 on GRCh38
- UniGene ID
- Hs.154704
- Species
-
Human