gRNA Design Details
- Target DNA Sequence
- AGATAGACTCAACAGGGCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 147831524 - 147831546 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK1
- Entrez Gene ID
-
6690
- Gene Name
- serine peptidase inhibitor, Kazal type 1
- Gene Aliases
-
PCTT, PSTI, Spink3, TATI, TCP
- Chromosome Location
-
Chr.5: 147824580 - 147839231 on GRCh38
- UniGene ID
- Hs.407856
- Species
-
Human