gRNA Design Details
- Target DNA Sequence
- GCGTGAAGGAGTATTTTGCC
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 69821067 - 69821089 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.83/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sptssb
- Entrez Gene ID
-
66183
- Gene Name
- serine palmitoyltransferase, small subunit B
- Gene Aliases
-
1110032A04Rik, ADMP, Sssptb
- Chromosome Location
-
Chr.3: 69819538 - 69859894 on GRCm38
- UniGene ID
- Mm.45481
- Species
-
Mouse