gRNA Design Details
- Target DNA Sequence
- GGCCCAATAACACACCACAA
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 47682087 - 47682109 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRF2
- Entrez Gene ID
-
222611
- Gene Name
- adhesion G protein-coupled receptor F2
- Gene Aliases
-
GPR111, PGR20, hGPCR35
- Chromosome Location
-
Chr.6: 47656590 - 47697797 on GRCh38
- UniGene ID
- Hs.724860
- Species
-
Human