gRNA Design Details
- Target DNA Sequence
- GCTCAGTAACGTGCTCGAAT
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 68956063 - 68956085 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRB3
- Entrez Gene ID
-
577
- Gene Name
- adhesion G protein-coupled receptor B3
- Gene Aliases
-
BAI3
- Chromosome Location
-
Chr.6: 68635282 - 69389511 on GRCh38
- UniGene ID
- Hs.13261
- Species
-
Human