gRNA Design Details
- Target DNA Sequence
- CACAATCCCTAGGACCTTTG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 231108976 - 231108998 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
96.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HTR2B
- Entrez Gene ID
-
3357
- Gene Name
- 5-hydroxytryptamine receptor 2B
- Gene Aliases
-
5-HT(2B), 5-HT-2B, 5-HT2B
- Chromosome Location
-
Chr.2: 231108230 - 231125778 on GRCh38
- UniGene ID
- Hs.421649
- Species
-
Human