gRNA Design Details
- Target DNA Sequence
- GCGGTTGAAGCGCAGGTAGA
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 31756582 - 31756604 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRB2
- Entrez Gene ID
-
576
- Gene Name
- adhesion G protein-coupled receptor B2
- Gene Aliases
-
BAI2
- Chromosome Location
-
Chr.1: 31727105 - 31764063 on GRCh38
- UniGene ID
- Hs.524138
- Species
-
Human