gRNA Design Details

Target DNA Sequence
AGCGTGACGTCTTCCTCCCA
PAM Sequence
GGG
Target locus
Chr.6: 46884156 - 46884178 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.97/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ADGRF5
Entrez Gene ID
221395
Gene Name
adhesion G protein-coupled receptor F5
Gene Aliases
GPR116, KPG_001
Chromosome Location
Chr.6: 46852505 - 46954954 on GRCh38
UniGene ID
Hs.362806
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001098518.1 NP_001091988.1 5