gRNA Design Details
- Target DNA Sequence
- AGCGTGACGTCTTCCTCCCA
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 46884156 - 46884178 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRF5
- Entrez Gene ID
-
221395
- Gene Name
- adhesion G protein-coupled receptor F5
- Gene Aliases
-
GPR116, KPG_001
- Chromosome Location
-
Chr.6: 46852505 - 46954954 on GRCh38
- UniGene ID
- Hs.362806
- Species
-
Human