gRNA Design Details
- Target DNA Sequence
- AGGCGCGGCGATGCTGTCGT
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 172447665 - 172447687 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GHSR
- Entrez Gene ID
-
2693
- Gene Name
- growth hormone secretagogue receptor
- Gene Aliases
-
GHDP
- Chromosome Location
-
Chr.3: 172443291 - 172448456 on GRCh38
- UniGene ID
- Hs.130212
- Species
-
Human