gRNA Design Details
- Target DNA Sequence
- CCGAAAACTGAGCGCCCACT
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 26346204 - 26346226 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRF3
- Entrez Gene ID
-
165082
- Gene Name
- adhesion G protein-coupled receptor F3
- Gene Aliases
-
GPR113, PGR23
- Chromosome Location
-
Chr.2: 26308173 - 26346817 on GRCh38
- UniGene ID
- Hs.631878
- Species
-
Human