gRNA Design Details

Target DNA Sequence
CGCTTCTCGCCCGCATGTCG
PAM Sequence
CGG
Target locus
Chr.7: 124764938 - 124764960 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 94/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GPR37
Entrez Gene ID
2861
Gene Name
G protein-coupled receptor 37
Gene Aliases
EDNRBL, PAELR, hET(B)R-LP
Chromosome Location
Chr.7: 124745601 - 124766025 on GRCh38
UniGene ID
Hs.406094
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_005302.3 NP_005293.1 1