gRNA Design Details
- Target DNA Sequence
- GTTCATACCTGAGGATCCGA
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 14396442 - 14396464 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
94.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRE5
- Entrez Gene ID
-
976
- Gene Name
- adhesion G protein-coupled receptor E5
- Gene Aliases
-
CD97, TM7LN1
- Chromosome Location
-
Chr.19: 14381144 - 14408725 on GRCh38
- UniGene ID
- Hs.466039
- Species
-
Human